Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17E/IL-25 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB12582100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB12582100 100 μg
MAB12582SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB12582100 Supplier R&D Systems Supplier No. MAB12582100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-17E/IL-25
Applications ELISA
Classification Monoclonal
Clone 878227
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q9H293
Gene Alias IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-17E/IL-25 Tyr33-Gly177 Accession # Q9H293
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 64806
Test Specificity Detects human IL-17E/IL-25 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.