Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17F Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC13351S100UG
This item is not returnable.
View return policy
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-17F | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96PD4 | |
| IL17F | |
| E. coli-derived recombinant human IL-17F Arg31-Gln163 Accession # Q96PD4 | |
| RUO | |
| 112744 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 197301 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-17F in direct ELISAs. Does not cross-react with recombinant human IL-17A, B, C, D, or E. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction