Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RA/IL-17R Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB177S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB177S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB177S100 Supplier R&D Systems Supplier No. FAB177S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RA/IL-17R
Applications Flow Cytometry
Classification Monoclonal
Clone 133617
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96F46
Gene Alias CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Gene Symbols IL17RA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17 RA/IL-17 R Leu33-Trp320 Accession # Q96F46
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Test Specificity Detects human IL-17 RA/IL-17 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-17 R, rmIL-17B R, or recombinant human IL-17B R is observed. In Western blots, no cross-reactivity with rmIL-17 R is observed
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.