Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RA/IL-17R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 13 publications
$214.00 - $2020.00
Specifications
Antigen | IL-17RA/IL-17R |
---|---|
Clone | 133617 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells |
Applications | Western Blot, Flow Cytometry, Neutralization, CyTOF, Immunoassay |
Description
IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, Knockout Validated.Specifications
IL-17RA/IL-17R | |
LYOPH | |
Western Blot, Flow Cytometry, Neutralization, CyTOF, Immunoassay | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
IL17RA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
133617 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization, CyTOF-ready, Knockout Validated 0.25 ug/10^6 cells | |
Monoclonal | |
Mouse | |
Human | |
Q96F46 | |
23765 | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 RA/IL-17 R Leu33-Trp320 Accession # Q96F46 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-17 RA/IL-17 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-17 R, rmIL-17B R, or recombinant human IL-17B R is observed. In Western blots, no cross-reactivity with rmIL-17 R is observed |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title