Learn More
Description
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA Development Kit |
| Quantity | 15 x 96-well Plates |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 23765 |
| Product Line | R&D Systems |
| Synonym | CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17 R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17 RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
