Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RA/IL-17R (Research Grade Brodalumab Biosimilar) Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB10547R10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB10547R10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB10547R10 Supplier R&D Systems Supplier No. FAB10547R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Antibody

IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Flow Cytometry

Specifications

Antigen IL-17RA/IL-17R
Applications Flow Cytometry
Classification Monoclonal
Clone Hu129
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A
Host Species Human
Immunogen Human Il-17 Receptor
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.