Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RC Alexa Fluor 488 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB22691G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB22691G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB22691G Supplier R&D Systems Supplier No. FAB22691G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17RC Monoclonal specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RC
Applications Flow Cytometry
Classification Monoclonal
Clone 309822
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_116121
Gene Alias FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14
Gene Symbols IL17RC
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17 RC Leu21-Ala454 Accession # NP_116121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84818
Test Specificity Detects human IL-17 RC in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 R, rhIL-17 BR, rhIL-17 RD, or recombinant mouse IL-17 RC is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.