Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RC Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB22691T100UG
This item is not returnable.
View return policy
Description
IL-17RC Monoclonal specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry.Specifications
IL-17RC | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_116121 | |
IL17RC | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 RC Leu21-Ala454 Accession # NP_116121 | |
RUO | |
84818 | |
Human | |
IgG2b |
Flow Cytometry | |
309822 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
Mouse | |
100 μg | |
Primary | |
Detects human IL-17 RC in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 R, rhIL-17 BR, rhIL-17 RD, or recombinant mouse IL-17 RC is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction