Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RC Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB22691
This item is not returnable.
View return policy
Description
IL-17RC Monoclonal specifically detects IL-17RC in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IL-17RC | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
NP_116121 | |
IL17RC | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 RC Leu21-Ala454 Accession # NP_116121 | |
100 μg | |
Primary | |
Detects human IL-17 RC in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-17 R, rhIL-17 BR, rhIL-17 RD, or recombinant mouse IL-17 RC is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
309822 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84818 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction