Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RD/SEF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | IL-17RD/SEF |
---|---|
Clone | 309539 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
IL-17RD/SEF Monoclonal specifically detects IL-17RD/SEF in Human samples. It is validated for Western Blot.Specifications
IL-17RD/SEF | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
AAM77571 | |
54756 | |
Mouse myeloma cell line NS0-derived recombinant human IL-17 RD/SEF Ala27-Arg299 Accession # AAM77571 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-17 RD/SEF in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse IL-17 RD, recombinant human (rh) IL-17 RC, or rhIL-17B R is observed. |
309539 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ35755, IL-17 RD, IL17RD, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLM, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, SEF, Sef homolog, SEFhSef | |
IL17RD | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title