Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RD/SEF PE-conjugated Antibody, R&D Systems™

Catalog No. FAB2275P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2275P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2275P Supplier R&D Systems Supplier No. FAB2275P
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-17RD/SEF Polyclonal specifically detects IL-17RD/SEF in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-17RD/SEF
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAM77571
Gene Alias FLJ35755, IL-17 RD, IL17RD, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLM, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, SEF, Sef homolog, SEFhSef
Gene Symbols IL17RD
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-17 RD/SEF Ala27-Arg299 Accession # AAM77571
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54756
Test Specificity Detects human IL-17 RD/SEF in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse (rm) IL-17 RD is observed and less than 10% cross-reactivity with recombinant human (rh) IL-17 RC, rmIL-17B R, and rhIL-17 R is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.