Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RE Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8358S100UG
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Flow Cytometry.Specifications
IL-17RE | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-17 RE Met1-His454 Accession # Q8NFR9 | |
RUO | |
132014 | |
Human | |
IgG1 |
Flow Cytometry | |
934832 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8NFR9 | |
Mouse | |
100 μg | |
Primary | |
Detects human IL-17 RE in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction