Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-17RE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8358
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8358 100 μg
MAB8358SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8358 Supplier R&D Systems Supplier No. MAB8358
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.

Specifications

Antigen IL-17RE
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Monoclonal
Clone 934832
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NFR9
Gene Alias IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E
Gene Symbols IL17RE
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IL-17 RE Met1-His454 Accession # Q8NFR9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014
Test Specificity Detects human IL-17 RE in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.