Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-17RE APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8358A025
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Flow Cytometry.Specifications
IL-17RE | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IL-17 RE, IL-17 receptor E, IL17RE, Il25r, interleukin 17 receptor E | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
132014 | |
Human | |
Purified |
Flow Cytometry | |
934832 | |
Flow Cytometry 10 uL/10^6 cells | |
Q8NFR9 | |
IL17RE | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-17 RE Met1-His454 Accession # Q8NFR9 | |
25 Tests | |
Primary | |
Detects human IL-17 RE in direct ELISAs. By flow cytometry, this antibody is specific for IL-17 RE and does not bind to cells transfected with IL-17 RA, IL-17 RB, or IL-17 RC. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction