Learn More
R&D Systems™ Human IL-18/IL-18 BPa Complex DuoSet ELISA
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Specifications
Specifications
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Binding Target | IL-18/IL-18 BPa Complex |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3606 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.