Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-18/IL-1F4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB91242SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB91242SP 25 μg
MAB91242100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB91242SP Supplier R&D Systems Supplier No. MAB91242SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA.

Specifications

Antigen IL-18/IL-1F4
Applications ELISA
Classification Monoclonal
Clone 884801
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q14116
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-18/IL-1F4 Tyr37-Asp193 Accession # Q14116
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity Detects human IL-18/IL-1F4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.