Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-18/IL-1F4 Propeptide Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB646
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB646 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB646 Supplier R&D Systems Supplier No. MAB646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated.

Specifications

Antigen IL-18/IL-1F4
Applications Western Blot, Flow Cytometry, CyTOF, Immunoassay
Classification Monoclonal
Clone 74801
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/10^6 cells, CyTOF-ready, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14116
Gene Alias Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
Gene Symbols IL18
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-18/IL-1F4 Accession # Q14116
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606
Test Specificity Detects the pro region of human IL-18/IL-1F4 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-18, recombinant mouse IL-18 or recombinant rat IL-18.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.