Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-18 R alpha/IL-1 R5 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB840T100UG
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry.Specifications
| IL-18R alpha/IL-1 R5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q13478 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects Recombinant Human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 70625 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IL18R1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478 | |
| RUO | |
| 8809 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction