Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-18 R alpha/IL-1 R5 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$233.50 - $502.50
Specifications
Antigen | IL-18R alpha/IL-1 R5 |
---|---|
Clone | 70625 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB840A025
![]() |
R&D Systems
FAB840A025 |
25 Tests |
Each for $233.50
|
|
|||||
FAB840A100
![]() |
R&D Systems
FAB840A100 |
100 Tests |
Each for $502.50
|
|
|||||
Description
IL-18R alpha/IL-1 R5 Monoclonal specifically detects IL-18R alpha/IL-1 R5 in Human samples. It is validated for Flow Cytometry.Specifications
IL-18R alpha/IL-1 R5 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
8809 | |
Mouse myeloma cell line NS0-derived recombinant human IL-18 R alpha/IL-1 R5 Glu20-Arg329 Accession # Q13478 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
70625 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q13478 | |
IL18R1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IL-18 R alpha/IL-1 R5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 R3, rhIL-1 R4, rhIL-1 R6, rhIL-1 R7, rhIL-1 R8, rhIL-1 R9, rhSIGIRR or recombinant mouse IL-18 R alpha is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title