Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-18 R alpha/IL-1 R5 ELISA Kit

Catalog No. EH263RB
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human IL-18 R alpha/IL-1 R5 quantitates human IL-18 R alpha/IL-1 R5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-18 R alpha/IL-1 R5.

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18 R-1; IL18RA; Il18ralpha; IL-18 R-alpha; IL18Ralpha2; Il1rrp; IL-1 Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha
Assay Range 28.67 to 7,000 pg/mL
Assay Sensitivity 28 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 8809
Gene Symbol Il18r1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL18R1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.