Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-18 R beta/IL-1 R7 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB118T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB118T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB118T100 Supplier R&D Systems Supplier No. FAB118T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-18 R beta/IL-1 R7/ACPL Monoclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-18 R beta/IL-1 R7/ACPL
Applications Flow Cytometry
Classification Monoclonal
Clone 132029
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O95256
Gene Alias Accessory protein-like, AcPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-1 R7, IL-18 receptor accessory protein, IL-18 receptor beta, IL18R beta, IL-18RAcP, IL18RAP, IL-18Rb, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
Gene Symbols IL18RAP
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-18 R beta/IL-1 R7 Met1-Arg356 Accession # O95256
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8807
Test Specificity Detects human IL-18 R beta/IL-1 R7 in direct ELISAs and Western blots. In direct ELISAs, 15% cross-reactivity with recombinant human (rh) IL-1 RII is observed and 2% cross-reactivity with rhIL-1 R9 is observed. Also in direct ELISAs, no cross-reactivity with rhIL-1 RI, rhIL-1 RacP/IL-1 R3, rhIL-1 R4, rhIL-18 R alpha/IL-1 R5, rhIL-1 R6, rhIL-1 R8, or recombinant mouse (rm) IL-18 R beta/IL-1 R7 is observed. In Western blots, approximately 100% cross-reactivity with rmIL-18 R beta is observed and no cross-reactivity with rhIL-18 R alpha/IL-1 R5, rhIL-1 RacP/IL-1 R3, rhIL-1 RI, rhIL-1 RII, or rhIL-1 Rrp2/IL-1 R6 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.