Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-18 R beta/IL-1 R7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF118SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF118SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF118SP Supplier R&D Systems Supplier No. AF118SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-18 R beta/IL-1 R7/ACPL Polyclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen IL-18 R beta/IL-1 R7/ACPL
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.3-1.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O95256
Gene Alias Accessory protein-like, AcPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-1 R7, IL-18 receptor accessory protein, IL-18 receptor beta, IL18R beta, IL-18RAcP, IL18RAP, IL-18Rb, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590
Gene Symbols IL18RAP
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-18 R beta/IL-1 R7 Phe20-Arg356 Accession # O95256
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8807
Test Specificity Detects human IL-18 R beta in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse IL-18 R beta (AcPL), recombinant human (rh) IL-18 R, rhIL-1 R1, rhIL-1 RII, and rhIL-1 Rrp2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.