Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-1ra/IL-1F3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF280NA
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF280NA 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF280NA Supplier R&D Systems Supplier No. AF280NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1ra/IL-1F3/IL1RN Polyclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization.

Specifications

Antigen IL-1ra/IL-1F3/IL1RN
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization ≤4 ug/mL
Gene Accession No. P18510
Gene Alias DIRA, ICIL-1RA, IL1F3, IL-1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4
Gene Symbols IL1RN
Host Species Goat
Immunogen E. coli-derived recombinant human IL-1ra/IL-1F3 Arg26-Glu177 Accession # P18510
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3557
Test Specificity Detects human IL-1ra/IL-1F3 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 15% cross-reactivity with recombinant mouse IL-1ra is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.