Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-1ra/IL-1F3 Quantikine ELISA Kit

Click to view available options
Quantity:
1 Plate
6 Plates
Description
The Human IL-1ra/IL-1F3 Quantikine ELISA Kit quantifies human IL-1ra/IL-1F3/IL1RN in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Assay Range | 31.20 - 2000pg/mL |
Assay Sensitivity | 18.3pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Specificity | Natural and Recombinant Human IL-1 ra |
Sufficient For | 1 x 96-well Plate |
Target Species | Human |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction