Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-1Rrp2/IL-1R6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
| Antigen | IL-1Rrp2/IL-1R6 |
|---|---|
| Clone | 116004 |
| Dilution | Neutralization 0.25-1.5 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB8721SP
![]() |
R&D Systems
MAB8721SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB8721100
![]() |
R&D Systems
MAB8721100 |
Unconjugated | 100 μg |
Each for $549.50
|
|
||||
Description
IL-1Rrp2/IL-1R6 Monoclonal specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Neutralization.Specifications
| IL-1Rrp2/IL-1R6 | |
| Neutralization 0.25-1.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-1 Rrp2/IL-1 R6 Asp20-Tyr337 Accession # Q9HB29 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-1 Rrp2/IL-1 R6 in direct ELISAs. |
| 116004 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HB29 | |
| 8808 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title