Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-2 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2023V100UG
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-2 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
E. coli-derived recombinant human IL-2, Ala21-Thr153, Accession # NP_000577 | |
100 μg | |
Primary | |
Detects human IL-2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
5310 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_000577 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3558 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction