Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $596.50
Specifications
Antigen | IL-2 |
---|---|
Clone | 5336R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2021SP
![]() |
R&D Systems
MAB2021SP |
25 μg |
Each for $241.00
|
|
|||||
MAB2021100
![]() |
R&D Systems
MAB2021100 |
100 μg |
Each for $596.50
|
|
|||||
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for ELISA.Specifications
IL-2 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
NP_000577 | |
3558 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
5336R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # NP_000577 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title