Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-2 R beta ELISA Kit (Colorimetric)
SDP

Catalog No. NB183414 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183414 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183414 Supplier Novus Biologicals™ Supplier No. NBP306790
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 78.13 to 5000 pg/mL
Assay Sensitivity 46.88 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Cell Culture Supernatant, Plasma, Serum
Specificity This kit recognizes Human IL-2 R beta in samples. No significant cross-reactivity or interference between Human IL-2 R beta and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3560
Gene Symbol IL2RB
Quantity 1 Kit
Research Discipline Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.