Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-20 R beta Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1788
This item is not returnable.
View return policy
Description
IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Human samples. It is validated for Western Blot.Specifications
IL-20 R beta/FNDC6 | |
Polyclonal | |
Biotin | |
Q6UXL0 | |
IL20RB | |
Mouse myeloma cell line NS0-derived recombinant human IL-20 R beta Asp30-Ala230 Accession # Q6UXL0 | |
50 μg | |
Primary | |
Detects human IL-20 R beta in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) IL-20 R alpha , rhIL-10 R beta , rhIFN-gamma R1, and rhIFN-gamma R2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FNDC6, DIRS1, FNDC6, IL20R beta, IL-20R2, IL-20Rb, IL20RB, IL20RB interleukin 20 receptor beta, MGC34923 | |
Goat | |
Affinity Purified | |
RUO | |
53833 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction