Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-20R alpha Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA11762U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA11762U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA11762U100 Supplier R&D Systems Supplier No. FAB11762U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-20R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 173714
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UHF4
Gene Alias class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20 R alpha, IL20R alpha, IL-20R1, IL-20R1IL-20 receptor subunit alpha, IL20RA, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7
Gene Symbols IL20RA
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-20 R alpha Val30-Lys250 Accession # Q9UHF4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53832
Test Specificity Detects human IL-20 R alpha in direct ELISAs and Western blots. In Western blots, approximately 5-15% cross-reactivity with recombinant human (rh) IFN-gamma R2, rhIL-10 R, rhIL-10 R beta , rhIL-20 R beta , and rhIL-22 BP is observed and no cross-reactivity with rhIFN-gamma RI, recombinant mouse IL-20 R alpha , or rhIL-22 R is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.