Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-20R alpha Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB11762N100UG
This item is not returnable.
View return policy
Description
IL-20R alpha Monoclonal specifically detects IL-20R alpha in Human samples. It is validated for Flow Cytometry.Specifications
IL-20R alpha | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9UHF4 | |
IL20RA | |
Mouse myeloma cell line NS0-derived recombinant human IL-20 R alpha Val30-Lys250 Accession # Q9UHF4 | |
RUO | |
53832 | |
Human | |
IgG1 |
Flow Cytometry | |
173714 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
class II cytokine receptor ZCYTOR7, CRF2-8, Cytokine receptor class-II member 8, Cytokine receptor family 2 member 8, FLJ40993, IL-20 R alpha, IL20R alpha, IL-20R1, IL-20R1IL-20 receptor subunit alpha, IL20RA, IL-20RA, IL-20R-alpha, interleukin 20 receptor, alpha, interleukin-20 receptor I, interleukin-20 receptor subunit alpha, ZcytoR7 | |
Mouse | |
100 μg | |
Primary | |
Detects human IL-20 R alpha in direct ELISAs and Western blots. In Western blots, approximately 5-15% cross-reactivity with recombinant human (rh) IFN-gamma R2, rhIL-10 R, rhIL-10 R beta , rhIL-20 R beta , and rhIL-22 BP is observed and no cross-reactivity with rhIFN-gamma RI, recombinant mouse IL-20 R alpha , or rhIL-22 R is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction