Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-21 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF15001100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF15001-100 100 μg
AF15001-SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF15001-100 Supplier R&D Systems Supplier No. AF15001100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Antibody

IL-21 specifically detects IL-21 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-21
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q9HBE4
Gene Alias CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11
Host Species Goat
Immunogen E. coli-derived recombinant human IL-21 Gln25-Ser155 Accession # Q9HBE4
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 59067
Test Specificity Detects human IL-21 in direct ELISAs.
Reconstitution Reconstitute at 0.2mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.