Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-21 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP184816 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP184816 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP184816 Supplier Novus Biologicals™ Supplier No. NBP184816
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human IL-21 ELISA Kit (Colorimetric) measures human IL-21 in cell culture supernatant, serum, plasma (edta, citrate).

Specifications

Assay Range 78 - 5,000pg/mL
Assay Sensitivity 20.0pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 59067
Gene Symbol IL21
Quantity 96 Tests
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform
Target IL-21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.