Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-21R Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9911R100UG
This item is not returnable.
View return policy
Description
IL-21R Monoclonal specifically detects IL-21R in Human samples. It is validated for Flow Cytometry.Specifications
| IL-21R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9HBE5 | |
| IL21R | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R Cys20-Pro236 Accession # Q9HBE5 | |
| RUO | |
| 50615 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 152512 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IL-21 R in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh)IL-2 R beta or rmIL-21 R. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction