Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-21R PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
$210.00 - $444.00
Specifications
Antigen | IL-21R |
---|---|
Clone | 152512 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB9911P025
![]() |
R&D Systems
FAB9911P025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB9911P100
![]() |
R&D Systems
FAB9911P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
IL-21R Monoclonal specifically detects IL-21R in Human samples. It is validated for Flow Cytometry.Specifications
IL-21R | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor | |
IL21R | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IL-21 R in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh)IL-2 R beta or rmIL-21 R. |
152512 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
Q9HBE5 | |
50615 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-21 R Cys20-Pro236 Accession # Q9HBE5 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title