Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7822R100
This item is not returnable.
View return policy
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-22 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
E. coli-derived recombinant human IL-22, Ala34-Ile179, Accession # Q9GZX6 | |
100 μg | |
50616 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG1 |
ELISA | |
142906R | |
ELISA Capture (Matched Antibody Pair) 2-8 μg/mL | |
Q9GZX6 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human IL-22 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction