Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$268.50 - $549.50
Specifications
Antigen | IL-22 |
---|---|
Clone | 142928 |
Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IC7821A025
![]() |
R&D Systems
IC7821A025 |
25 Tests |
Each for $268.50
|
|
|||||
IC7821A100
![]() |
R&D Systems
IC7821A100 |
100 Tests |
Each for $549.50
|
|
|||||
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-22 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
IL22 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. |
142928 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q9GZX6 | |
50616 | |
E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title