Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22 R alpha 1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2770R100UG
This item is not returnable.
View return policy
Description
IL-22R alpha 1 Monoclonal specifically detects IL-22R alpha 1 in Human samples. It is validated for Flow Cytometry.Specifications
IL-22R alpha 1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8N6P7 | |
IL22RA1 | |
Chinese hamster ovary cell line CHO-derived recombinant human IL-22 R alpha 1 Pro18-Thr228 Accession # Q8N6P7 | |
RUO | |
58985 | |
Human | |
IgG1 |
Flow Cytometry | |
305405 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CRF2-9, CRF2-9interleukin 22 receptor, Cytokine receptor class-II member 9, Cytokine receptor family 2 member 9, IL-22 receptor subunit alpha-1, IL22R, IL22R alpha 1, IL22R1, IL22RA1, IL-22RA1, IL-22R-alpha-1, IL-TIF-R1, interleukin 22 receptor, alpha 1, interleukin-22 receptor subunit alpha-1, zcytoR11 | |
Mouse | |
100 μg | |
Primary | |
Detects human IL-22 R alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-10, rhIL-22BP or rhIL-20 R alpha is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction