Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-22BP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $561.50
Specifications
Antigen | IL-22BP/IL22 RA2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-22BP/IL22 RA2 Polyclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.Specifications
IL-22BP/IL22 RA2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16 | |
IL22RA2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_851826 | |
116379 | |
Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro231 Accession # NP_851826 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IL-22BP in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human IL-22 R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title