Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-22BP APC-conjugated Antibody, R&D Systems™

Catalog No. IC10871A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC10871A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC10871A025 Supplier R&D Systems Supplier No. IC10871A025
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-22BP/IL22 RA2 Monoclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-22BP/IL22 RA2
Applications Flow Cytometry
Classification Monoclonal
Clone 875504
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q969J5
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Gene Symbols IL22RA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro263 Accession # Q969J5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116379
Test Specificity Detects human IL-22BP in direct ELISAs. In direct ELISAs, no cross-reactivity with mouse IL-22BP is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.