Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-22BP Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1087
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1087 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1087 Supplier R&D Systems Supplier No. BAF1087
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-22BP/IL22 RA2 Polyclonal specifically detects IL-22BP/IL22 RA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen IL-22BP/IL22 RA2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. NP_851826
Gene Alias class II cytokine receptor, CRF2-10, CRF2-S1IL22BP, CRF2-X, Cytokine receptor class-II member 10, Cytokine receptor family 2 member 10, Cytokine receptor family type 2, soluble 1, IL-22 RA2, IL-22 receptor subunit alpha-2, IL22BP, IL-22BPCRF2X, IL22RA2, IL-22RA2, IL-22R-alpha-2, interleukin 22 receptor, alpha 2, interleukin 22-binding protein, interleukin-22 receptor subunit alpha-2, Interleukin-22-binding protein, MGC150509, MGC150510, zcytoR16
Gene Symbols IL22RA2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-22BP Thr22-Pro231 Accession # NP_851826
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116379
Test Specificity Detects human IL-22BP in Western blots. In this format, approximately 5% cross-reactivity with recombinant human IL-22 R is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.