Learn More
Gibco™ Human IL-23 Recombinant Protein

Description
Carrier-Free Reconstitution: Lyophilized human IL-23 should be reconstituted in water to a protein concentration of 0.1-1.0 mg/mL. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -80°C to -20°C. Storage: Lyophilized human IL-23 should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human IL-23 at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

Specifications
Specifications
Accession Number | Q9NPF7 |
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 51561 |
Molecular Weight (g/mol) | 55 kDa |
Name | Human IL-23 |
Purification Method | Sequential Chromatography |
Quantity | 10 μg |
Endotoxin Concentration | <0.1 ng/ μg |
Gene Alias | il 23; IL 23 A; IL12B; Il-12 b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23 A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12 B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798 |
Show More |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.