Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-23R PerCP-conjugated Antibody, R&D Systems™

Catalog No. FAB14001C10
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB14001C10 100 Tests
FAB14001C02 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB14001C10 Supplier R&D Systems Supplier No. FAB14001C100
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-23R Monoclonal specifically detects IL-23R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-23R
Applications Flow Cytometry
Classification Monoclonal
Clone 218213
Conjugate PerCP
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q5VWK5
Gene Alias IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor
Gene Symbols IL23R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-23 R Gly24-Ile354 Accession # Q5VWK5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149233
Test Specificity Detects human IL-23 R in direct ELISAs and Western blots. Does not cross-react with recombinant mouse IL-23 R.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.