Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-26/AK155 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC13751U100UG
This item is not returnable.
View return policy
Description
IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-26/AK155 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NPH9.1 | |
IL26 | |
E. coli-derived recombinant human IL-26/AK155 Lys22-Gln171 Accession # Q9NPH9.1 | |
RUO | |
55801 | |
Human | |
IgG1 |
Flow Cytometry | |
510414 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155 | |
Mouse | |
100 μg | |
Primary | |
Detects human IL-26/AK155 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction