Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-26/AK155 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 4 publications

$200.50 - $2020.00

Specifications

Antigen IL-26/AK155
Clone 197505
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1375SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1375SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB1375100
GREENER_CHOICE
View Documents
R&D Systems
MAB1375100
100 μg
Each for $538.00
Only null left
Add to Cart
 
MAB1375500
GREENER_CHOICE
View Documents
R&D Systems
MAB1375500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

IL-26/AK155 Monoclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-26/AK155
LYOPH
Western Blot
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155
IL26
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
197505
Western Blot 1 ug/mL
Monoclonal
Mouse
Human
Q9NPH9
55801
E. coli-derived recombinant human IL-26 Lys22-Gln171 Accession # Q9NPH9
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IL-26 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human IL-10.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.