Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human IL-26/AK155 Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $615.44
Specifications
| Antigen | IL-26/AK155 | 
|---|---|
| Concentration | LYOPH | 
| Dilution | Western Blot 0.1 ug/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
Description
IL-26/AK155 Polyclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.Specifications
| IL-26/AK155 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155 | |
| IL26 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NPH9 | |
| 55801 | |
| E. coli-derived recombinant human IL-26 (R&D Systems, Catalog # 1375-IL) Lys22-Gln171 Accession # Q9NPH9 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human IL-26 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human IL-10 is observed. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title