Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-27 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems BAF2526
This item is not returnable.
View return policy
Description
IL-27 Polyclonal specifically detects IL-27 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
IL-27 | |
Polyclonal | |
Biotin | |
Q14213 (EBI-3 subunit), AAM34498 (p28 subunit) | |
IL27 | |
Mouse myeloma cell line NS0-derived recombinant human IL-27 (R&D Systems, Catalog # 2526-IL) Arg21-Lys229 (IL-27 EBI-3 subunit), Phe29-Pro243 (IL-27 p28 subunit) Accession # Q14213 (EBI-3 subunit), AAM34498 (p28 subunit) | |
50 μg | |
Primary | |
Detects human IL-27 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant mouse IL-27, recombinant human (rh) IL-12, and rhIL-12 p40 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A | |
Goat | |
Affinity Purified | |
RUO | |
246778 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction