Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-27/IL-35 EBI3 Subunit Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6456R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6456R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6456R100 Supplier R&D Systems Supplier No. IC6456R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-27/IL-35 EBI3 Subunit
Applications Flow Cytometry
Classification Monoclonal
Clone 607201
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q14213
Gene Alias cytokine receptor, EBI-3, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 beta, IL-27 subunit beta, IL27B, IL-27B, IL35 EBI3, IL-35 EBI3, interleukin-27 subunit beta
Gene Symbols EBI3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-27/IL-35 EBI3 Subunit Arg21-Lys229 (predicted) Accession # Q14213
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10148
Test Specificity Detects human IL-27/IL-35 EBI3 Subunit in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-27 EBI3 or rmIL-27 (EBI3 + p28 heterodimer) is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.