Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-27 PE-conjugated Antibody, R&D Systems™

Catalog No. IC25261P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC25261P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC25261P Supplier R&D Systems Supplier No. IC25261P
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-27 Monoclonal specifically detects IL-27 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-27
Applications Flow Cytometry
Classification Monoclonal
Clone 307426
Conjugate R-PE
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias IL27, IL-27, IL-27 p28 subunit, IL-27 subunit alpha, IL27A, IL-27-A, IL27-A, IL27p28, IL30, interleukin 27, interleukin 30, interleukin-27 subunit alpha, MGC71873, p28IL-27A
Gene Symbols IL27
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-27 Arg21-Lys229 of EBI-3 (Accession #Q14213.2) and Phe29-Pro243 of p28 (Accession #AAM34498)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246778
Test Specificity Detects human IL-27 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.