Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IL-28A/IFN-lambda 2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB15871500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB15871500 500 μg
MAB15871SP 25 μg
MAB15871100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB15871500 Supplier R&D Systems Supplier No. MAB15871500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-28A/IFN-lambda 2 Monoclonal specifically detects IL-28A/IFN-lambda 2 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-28A/IFN-lambda 2
Applications ELISA
Classification Monoclonal
Clone 248512
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IZJ0
Gene Alias interleukin-28A, IFN-lambda 2, IL28A, IL-28A, interferon, lambda 2
Gene Symbols IFNL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-28A/IFN-lambda 2 Val26-Val200 Accession # Q8IZJ0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282616
Test Specificity Detects human IL-28A/IFN-lambda 2 in ELISAs. In sandwich immunoassays, 3.5% cross-reactivity with human IL-28B/IFN-lambda 3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.